Anti-ELYS Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-9880R-A680
BOSSBS-9880R-A680EA 893 EUR
BOSSBS-9880R-A680
Anti-ELYS Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ELYS is a protein of the nuclear matrix that contains one AT hook DNA-binding domain. ELYS is a dual nucleoporin/kinetochore protein required for nuclear pore assembly and proper cell division. Nuclear pores span the nuclear envelope and act as gated aqueous channels to regulate the transport of macromolecules between the nucleus and cytoplasm, from individual proteins and RNAs to entire viral genomes. The A.T. hook near the C-terminus of likely indicates ELYS can function as a transcription factor which may play a specific role in hematopoietic tissues. It also may play a role in early embryo development, as ELYS is critical for the survival of inner cells of the blastocyst.

Type: Primary
Antigen: ELYS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 451-550/2266
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR