Anti-GLCNE Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-9882R-A680
BOSSBS-9882R-A680EA 893 EUR
BOSSBS-9882R-A680
Anti-GLCNE Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The bifunctional enzyme UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase (GNE/Mnk), or GLCNE, regulates and initiates biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. GLCNE is required for normal sialylation in hematopoietic cells. Sialylation is implicated in cell adhesion, signal transduction, tumourigenicity and metastatic behavior of malignant cells. It is upregulated after PKC-dependent phosphorylation and is most abundantly expressed in liver and placenta. It is also expressed, to a lesser extent, in heart, brain, lung, kidney, skeletal muscle and pancreas. Defects in GLCNE are the cause of sialuria, inclusion body myopathy 2 (IBM2) and Nonaka myopathy (NM) or distal myopathy with rimmed vacuoles (DMRV). Sialuria is an autosomal dominant disorder caused by a lack of feedback inhibition of GLCNE by CMP-NeuAc, resulting in overproduction of NeuAc. It is characterised by an accumulation of free sialic acid in the cytoplasm and large quantities of neuraminic acid in the urine. Both IBM2 and NM/DMRV are autosomal recessive neuromuscular disorders characterised by adult onset, distal and proximal muscle weakness (especially in the legs) and a typical muscle pathology including filamentous inclusions and rimmed vacuoles.

Type: Primary
Antigen: GLCNE
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 131-230/722
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR