Anti-GGT6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-9909R-A680
BOSSBS-9909R-A680EA 893 EUR
BOSSBS-9909R-A680
Anti-GGT6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
GGT6 belongs to the gamma-glutamyltransferase family. GGT is a membrane-bound extracellular enzyme that cleaves gamma-glutamyl peptide bonds in glutathione and other peptides and transfers the gamma-glutamyl moiety to acceptors. GGT is also key to glutathione homeostasis because it provides substrates for glutathione synthesis. GGT6 has very limited amino acid similarity to GGT1 and it's enzymatic activities are currently uncharacterised. GGT assays are of current widespread clinical use to help assess tissue damage.

Type: Primary
Antigen: GGT6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 321-420/493
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR