Anti-DGCR6L Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-9990R-A680
BOSSBS-9990R-A680EA 893 EUR
BOSSBS-9990R-A680
Anti-DGCR6L Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Neural crest cell migration to the third and fourth pharyngeal pouches is a critical step in the structural formation of organs that are affected in DiGeorge syndrome. DGCR6 (DiGeorge syndrome critical region 6) is a nuclear protein that plays a role in neural crest cell migration and is located at the DiGeorge syndrome critical region (DGCR) on chromosome 22. Expressed ubiquitously with highest levels in heart, liver and skeletal muscle, DGCR6 shares high homology with the Drosophila gonadal (gdl) protein and with human Laminin-1, both of which are involved in early tissue development. The gene encoding DGCR6, along with other DGCR genes, is deleted in DiGeorge syndrome; a developmental disorder characterised by improper facial, cardiac and palate formation. Upregulation of DGCR6 is implicated in lung and colon adenocarcinomas, as well as in Burkitt's lymphoma and lymphocytes transformed by EBV. Due to a duplication of the ancestral DGCR6 locus, there are two functional, highly homologous copies of the DGCR6 gene (designated DGCR6 and DGCR6L) on chromosome 22.

Type: Primary
Antigen: DGCR6L
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-130/220
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR