PSA Mouse Monoclonal Antibody [clone: Pa3] (ALEXA FLUOR® 680)
Lieferant: Bioss
BSM-1625M-A680
BOSSBSM-1625M-A680EA 1080 EUR
BOSSBSM-1625M-A680
PSA Mouse Monoclonal Antibody [clone: Pa3] (ALEXA FLUOR® 680)
Antikörper
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its protein product is a protease present in seminal plasma. It is thought to function normally in the liquefaction of seminal coagulum, presumably by hydrolysis of the high molecular mass seminal vesicle protein. Serum level of this protein, called PSA in theMore Product Information