Anti-AKT1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00024
BSBTA00024EA 0 EUR
BSBTA00024
Anti-AKT1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for RAC-alpha serine/threonine-protein kinase(Akt1) detection. Tested with WB in Mouse, Rat.

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR