Anti-PD-L1/B7-H1 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
A00109
BSBTA00109EA 552 EUR
BSBTA00109
Anti-PD-L1/B7-H1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Programmed cell death 1 ligand 1(CD274) detection. Tested with WB in Human.
Type: Primary
Antigen: PD-L1/B7-H1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human