Anti-IL33 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00113
BSBTA00113EA 552 EUR
BSBTA00113
Anti-IL33 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for IL33 detection. Tested with WB, IHC-P, ELISA(Cap) in Human.

Type: Primary
Antigen: IL33
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR