Anti-MPI Rabbit Polyclonal Antibody

Lieferant: BosterBio
A00175
BSBTA00175EA 552 EUR
BSBTA00175
Anti-MPI Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Mannose-6-phosphate isomerase(MPI) detection. Tested with WB in Human, Rat.

Type: Primary
Antigen: MPI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR