Anti-Flt3 / CD135 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
A00188-3
BSBTA00188-3EA 552 EUR
BSBTA00188-3
Anti-Flt3 / CD135 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Flt3 / CD135 detection. Tested with WB, IHC-P, ELISA(Cap) in Human.
Type: Primary
Antigen: Flt3 / CD135
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human