Anti-KCNQ1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00310-1
BSBTA00310-1EA 552 EUR
BSBTA00310-1
Anti-KCNQ1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Potassium voltage-gated channel subfamily KQT member 1(KCNQ1) detection. Tested with WB in Human, Rat.

Type: Primary
Antigen: KCNQ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR