Anti-ATF4 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00371
BSBTA00371EA 552 EUR
BSBTA00371
Anti-ATF4 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cyclic AMP-dependent transcription factor ATF-4(ATF4) detection. Tested with WB in Human.

Type: Primary
Antigen: ATF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR