Anti-GST3 / GST pi Rabbit Polyclonal Antibody
Lieferant: Boster Bio
A00394-1
BSBTA00394-1EA 552 EUR
BSBTA00394-1
Anti-GST3 / GST pi Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for GST3 / GST pi detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: GST3 / GST pi
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat