Anti-KRT5 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00398
BSBTA00398EA 552 EUR
BSBTA00398
Anti-KRT5 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Keratin, type II cytoskeletal 5(KRT5) detection. Tested with WB, IHC-P in Human, Mouse, Rat.

Type: Primary
Antigen: KRT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR