Anti-SPR Polyclonal Antibody

Lieferant: Boster Bio
A00416-1
BSBTA00416-1EA 552 EUR
BSBTA00416-1
Anti-SPR Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for SPR detection. Tested with WB, Direct ELISA in Human;Mouse.

Type: Primary
Antigen: SPR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR