Anti-Topoisomerase I Rabbit Polyclonal Antibody
Lieferant: Boster Bio
A00434-1
BSBTA00434-1EA 552 EUR
BSBTA00434-1
Anti-Topoisomerase I Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Topoisomerase I detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: Topoisomerase I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat