Anti-Galectin 1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00470-1
BSBTA00470-1EA 552 EUR
BSBTA00470-1
Anti-Galectin 1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Galectin 1 detection. Tested with WB, IHC-P, ELISA(Cap) in Human;Mouse;Rat.

Type: Primary
Antigen: Galectin 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR