Anti-TIMP1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00561
BSBTA00561EA 552 EUR
BSBTA00561
Anti-TIMP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for TIMP1 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: TIMP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR