Anti-Ogg1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00768-1
BSBTA00768-1EA 552 EUR
BSBTA00768-1
Anti-Ogg1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Ogg1 detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: Ogg1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR