Anti-TLS / FUS Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00771-1
BSBTA00771-1EA 552 EUR
BSBTA00771-1
Anti-TLS / FUS Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for TLS / FUS detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: TLS / FUS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR