Anti-CD59 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00914
BSBTA00914EA 552 EUR
BSBTA00914
Anti-CD59 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for CD59 detection. Tested with WB, IHC-P, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: CD59
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR