Anti-Follistatin Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A00972-1
BSBTA00972-1EA 552 EUR
BSBTA00972-1
Anti-Follistatin Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Follistatin detection. Tested with WB, ELISA(Cap) in Human;Mouse;Rat.

Type: Primary
Antigen: Follistatin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR