Anti-BCMA Polyclonal Antibody

Lieferant: BosterBio
A01014-1
BSBTA01014-1EA 552 EUR
BSBTA01014-1
Anti-BCMA Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for BCMA detection. Tested with WB, IHC-P, ELISA(Cap) in Human.

Type: Primary
Antigen: BCMA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR