Anti-14-3-3 sigma Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A01127
BSBTA01127EA 552 EUR
BSBTA01127
Anti-14-3-3 sigma Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for 14-3-3 sigma detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: 14-3-3 sigma
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR