Anti-IL12B Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A01152-3
BSBTA01152-3EA 552 EUR
BSBTA01152-3
Anti-IL12B Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for IL12B detection. Tested with WB, IHC-P, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: IL12B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR