Anti-PPIA Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A01308
BSBTA01308EA 552 EUR
BSBTA01308
Anti-PPIA Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Peptidyl-prolyl cis-trans isomerase A(PPIA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: PPIA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR