Anti-CD200/OX2 Polyclonal Antibody

Lieferant: BosterBio
A01512-1
BSBTA01512-1EA 552 EUR
BSBTA01512-1
Anti-CD200/OX2 Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for CD200/OX2 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: CD200/OX2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR