Anti-CKB Polyclonal Antibody

Lieferant: Boster Bio
A01695-1
BSBTA01695-1EA 552 EUR
BSBTA01695-1
Anti-CKB Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for CKB detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: CKB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR