Anti-RUNX1T1/ETO Polyclonal Antibody

Lieferant: Boster Bio
A01792-1
BSBTA01792-1EA 552 EUR
BSBTA01792-1
Anti-RUNX1T1/ETO Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for RUNX1T1/ETO detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: RUNX1T1/ETO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR