Anti-PER3 Rabbit Polyclonal Antibody
Lieferant: Boster Bio
A01835-1
BSBTA01835-1EA 552 EUR
BSBTA01835-1
Anti-PER3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for PER3 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: PER3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat