Anti-ANGPTL8 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A02471
BSBTA02471EA 552 EUR
BSBTA02471
Anti-ANGPTL8 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Angiopoietin-like protein 8 (ANGPTL8) detection. Tested with WB in Human.

Type: Primary
Antigen: ANGPTL8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR