Anti-BAFF Receptor Polyclonal Antibody

Lieferant: Boster Bio
A02865-2
BSBTA02865-2EA 552 EUR
BSBTA02865-2
Anti-BAFF Receptor Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for BAFF Receptor detection. Tested with WB, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: BAFF Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR