Anti-Caspase 4 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A02941-1
BSBTA02941-1EA 552 EUR
BSBTA02941-1
Anti-Caspase 4 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Caspase 4 detection. Tested with WB, IHC-P, Direct ELISA in Human.

Type: Primary
Antigen: Caspase 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR