Anti-ARFGAP1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A04959
BSBTA04959EA 552 EUR
BSBTA04959
Anti-ARFGAP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for ADP-ribosylation factor GTPase-activating protein 1(ARFGAP1) detection. Tested with WB in Human, Mouse, Rat.

Type: Primary
Antigen: ARFGAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR