Anti-HnRNP F Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A05806
BSBTA05806EA 552 EUR
BSBTA05806
Anti-HnRNP F Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Heterogeneous nuclear ribonucleoprotein F(HNRNPF) detection. Tested with WB, IHC-P in Human, Mouse, Rat.

Type: Primary
Antigen: HnRNP F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR