Anti-FMN1 Rabbit Polyclonal Antibody

Lieferant: Boster Bio
A07520
BSBTA07520EA 552 EUR
BSBTA07520
Anti-FMN1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Formin-1(FMN1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: FMN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR