Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Lieferant: BosterBio
BA1050-1
BSBTBA1050-1EA 0 EUR
BSBTBA1050-1
Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity:
Bestellen