Anti-IgG Rabbit Antibody (HRP (Horseradish Peroxidase))

Lieferant: BosterBio
BA1070-1
BSBTBA1070-1EA 0 EUR
BSBTBA1070-1
Anti-IgG Rabbit Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR