Anti-IgG Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Boster Bio
BA1089-0.5
BSBTBA1089-0.5EA 0 EUR
BSBTBA1089-0.5
Anti-IgG Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR