Anti-IgG Goat Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Lieferant: BosterBio
BA1090-0.5
BSBTBA1090-0.5EA 0 EUR
BSBTBA1090-0.5
Anti-IgG Goat Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen