Anti-Protein A Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Boster Bio
BA1120-0.5
BSBTBA1120-0.5EA 0 EUR
BSBTBA1120-0.5
Anti-Protein A Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Tag
Antigen: Protein A
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR