Anti-IgG Mouse Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Boster Bio
BM2003-0.5
BSBTBM2003-0.5EA 0 EUR
BSBTBM2003-0.5
Anti-IgG Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR