Anti-Gelsolin Mouse Monoclonal Antibody [clone: GEL-42]
Lieferant: Boster Bio
MA1044
BSBTMA1044EA 552 EUR
BSBTMA1044
Anti-Gelsolin Mouse Monoclonal Antibody [clone: GEL-42]
Antikörper
Mouse IgG monoclonal antibody for Gelsolin, gelsolin (GSN) detection. Tested with WB, IHC-P, IHC-F in Human; rabbit; bovine. No cross reactivity with otherproteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Gelsolin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit