Anti-MAP2 Mouse Monoclonal Antibody [clone: MP-2]
Lieferant: Boster Bio
MA1057
BSBTMA1057EA 552 EUR
BSBTMA1057
Anti-MAP2 Mouse Monoclonal Antibody [clone: MP-2]
Antikörper
Mouse IgG monoclonal antibody for MAP2, microtubule-associated protein 2 (MAP2) detection. Tested with WB, IHC-P in Human;mouse;rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse