Anti-PCK Mouse Monoclonal Antibody [clone: IML-91]
Lieferant: Boster Bio
MA1081
BSBTMA1081EA 552 EUR
BSBTMA1081
Anti-PCK Mouse Monoclonal Antibody [clone: IML-91]
Antikörper
Mouse IgG monoclonal antibody for PCK detection. Tested with WB, IHC-P, IHC-F in Human; rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: PCK
Clonality: Monoclonal
Clone: IML-91
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a/ IgG1
Reactivity: Human, Rat