Anti-Vinculin Mouse Monoclonal Antibody [clone: VIN-54]

Lieferant: Boster Bio
MA1103
BSBTMA1103EA 572 EUR
BSBTMA1103
Anti-Vinculin Mouse Monoclonal Antibody [clone: VIN-54]
Antikörper
Mouse IgG monoclonal antibody for Vinculin, vinculin (VCL) detection. Tested with WB, IHC-F in Human;mouse;rat;chicken. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Vinculin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR