Anti-alpha-Tubulin Mouse Monoclonal Antibody [clone: Tub-1]
Lieferant: Boster Bio
MA1107
BSBTMA1107EA 552 EUR
BSBTMA1107
Anti-alpha-Tubulin Mouse Monoclonal Antibody [clone: Tub-1]
Antikörper
Mouse IgG monoclonal antibody for alpha-Tubulin detection. Tested with WB, IHC-P in Human;rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: alpha-Tubulin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken