Anti-MIF (macrophage migration inhibitory factor (glycosylation-inhibiting factor)) Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1052
BSBTPA1052EA 552 EUR
BSBTPA1052
Anti-MIF (macrophage migration inhibitory factor (glycosylation-inhibiting factor)) Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Macrophage migration inhibitory factor(MIF) detection. Tested with WB,IHC-P in Human;Mouse;Rat.

Macrophage migration inhibitory factor, MIF, is a cytokine released by T-lymphocytes, macrophages, and the pituitary gland that serves to integrate peripheral and central inflammatory responses. MIF gene has 3 exons separated by introns of only 189 and 95 bp, and covers less than 1 kb. Localization of the human gene for macrophage migration inhibitory factor(MIF) to chromosome 22q11.2 MIF plays a critical role in inflammatory diseases and atherogenesis.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: MIF (macrophage migration inhibitory factor (glycosylation-inhibiting factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR