Anti-RAGE Rabbit Polyclonal Antibody

Lieferant: Boster Bio
PA1069
BSBTPA1069EA 552 EUR
BSBTPA1069
Anti-RAGE Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Advanced glycosylation end product-specific receptor (AGER) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: RAGE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR