Anti-GFAP Rabbit Polyclonal Antibody

Lieferant: BosterBio
PA1239
BSBTPA1239EA 552 EUR
BSBTPA1239
Anti-GFAP Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Glial fibrillary acidic protein (GFAP) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR